Order Entry
Italy
ContactUsLinkComponent
Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-48]
Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-48]
Catalog # USBII7661-60
Supplier:  US Biological
Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-48]
Catalog # USBII7661-60
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interferon alpha 1
  • Antigen symbol:
    IFNA1
  • Clonality:
    Monoclonal
  • Clone:
    2-48
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Interferon alpha 1
Clonality: monoclonal
Clone: 17564
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human