Anti-ILT3 Mouse Monoclonal Antibody [clone: ZM4.1]
Catalog # USBII7502-75E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin T3
- Antigen symbol:ILT3
- Clonality:Monoclonal
- Clone:ZM4.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ILT3
Clonality: monoclonal
Clone: ZM4.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human