Order Entry
Italy
ContactUsLinkComponent
 
Anti-Hydroxysteroid Dehydrogenase, 17-beta 7 Rabbit Polyclonal Antibody
Catalog # USBIH9117-83T
Supplier:  US Biological
undefined
Anti-Hydroxysteroid Dehydrogenase, 17-beta 7 Rabbit Polyclonal Antibody
Catalog # USBIH9117-83T
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Hydroxysteroid Dehydrogenase, 17-beta 7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: Hydroxysteroid Dehydrogenase, 17-beta 7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human