Anti-17a-Hydroxyprogesterone-3-CMO Sheep Polyclonal Antibody
Catalog # USBIH9111-32R
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:17-alpha Hydroxyprogesterone-3-CMO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: 17a-Hydroxyprogesterone-3-CMO
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human