Order Entry
Italy
ContactUsLinkComponent
Anti-HLA Class 2 Antigen DQ1, DQ3 Mouse Monoclonal Antibody [clone: HL-37]
Anti-HLA Class 2 Antigen DQ1, DQ3 Mouse Monoclonal Antibody [clone: HL-37]
Catalog # USBIH6099-94T
Supplier:  US Biological
Anti-HLA Class 2 Antigen DQ1, DQ3 Mouse Monoclonal Antibody [clone: HL-37]
Catalog # USBIH6099-94T
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 2 Antigen DQ1, DQ3
  • Clonality:
    Monoclonal
  • Clone:
    HL-37
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG3
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HLA Class 2 Antigen DQ1, DQ3
Clonality: monoclonal
Clone: HL-37
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: