Anti-HLA Class 2 Antigen D Mouse Monoclonal Antibody [clone: 7.5.10.1]
Catalog # USBIH6099-81B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen D
- Clonality:Monoclonal
- Clone:7.5.10.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:5
Type: Primary
Antigen: HLA Class 2 Antigen D
Clonality: monoclonal
Clone: 7.5.10.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human