Anti-HLA Class 1 Antigen E Rabbit Polyclonal Antibody (Biotin)
USBIH6098-68B-BIOT
- Antibody type:Primary
- Antigen name:HLA Class 1 Antigen E
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: HLA Class 1 Antigen E
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human