Order Entry
Italy
ContactUsLinkComponent
Anti-HLA Class 1 Antigen ABC Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: W6/32]
Anti-HLA Class 1 Antigen ABC Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: W6/32]
Catalog # USBIH6098-60D
Supplier:  US Biological
Anti-HLA Class 1 Antigen ABC Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: W6/32]
Catalog # USBIH6098-60D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 1 Antigen ABC
  • Clonality:
    Monoclonal
  • Clone:
    W6/32
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    saline, 0,5% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,025 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: HLA Class 1 Antigen ABC
Clonality: monoclonal
Clone: W6/32
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human