Order Entry
Italy
ContactUsLinkComponent
 
Anti-Histone H3, Citrulline 2, 8, 17 Rabbit polyclonal antibody
Catalog # USBIH5110-12Q
Supplier:  US Biological
Anti-Histone H3, Citrulline 2, 8, 17 Rabbit polyclonal antibody
Catalog # USBIH5110-12Q
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Histone H3, Citrulline 2, 8, 17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage buffer:
    200ul ddH2O.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:200-1:1000

Type: Primary
Antigen: Histone H3, Citrulline 2, 8, 17
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human