Anti-Hepatitis C virus Core, NS3, NS4 Goat Polyclonal Antibody
Catalog # USBIH1920-13C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis C virus Core, NS3, NS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~4-5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Hepatitis C virus Core, NS3, NS4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: