Order Entry
Italy
ContactUsLinkComponent
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: 51-13]
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: 51-13]
Catalog # USBIH1840-01B
Supplier:  US Biological
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: 51-13]
Catalog # USBIH1840-01B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Helicobacter pylori
  • Clonality:
    Monoclonal
  • Clone:
    51-13
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Form:
    liquid
  • Storage buffer:
    PBS, 1% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: Helicobacter pylori
Clonality: monoclonal
Clone: 51-13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: