Order Entry
Italy
ContactUsLinkComponent
Anti-H Type 2 Antigen Mouse Monoclonal Antibody [clone: 19-OLE]
Anti-H Type 2 Antigen Mouse Monoclonal Antibody [clone: 19-OLE]
Catalog # USBIH0010-20
Supplier:  US Biological
Anti-H Type 2 Antigen Mouse Monoclonal Antibody [clone: 19-OLE]
Catalog # USBIH0010-20
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    H Type 2 Antigen
  • Clonality:
    Monoclonal
  • Clone:
    19-OLE
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: H Type 2 Antigen
Clonality: monoclonal
Clone: 19-OLE
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human