Anti-pro-Guanylin Mouse Monoclonal Antibody [clone: MAK-L-G-11]
Catalog # USBIG9805-04E-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:pro-Guanylin
- Clonality:Monoclonal
- Clone:MAK-L-G-11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:lyophilized powder
- Amino acid number:78 to 94
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:10 µg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: pro-Guanylin
Clonality: monoclonal
Clone: MAK-L-G-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: