Anti-Follicle Stimulating Hormone beta 2 Mouse monoclonal antibody [clone: INN-hFSH-60]
Catalog # USBIF5900-21C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Follicle Stimulating Hormone beta 2
- Antigen symbol:FSHB2
- Clonality:Monoclonal
- Clone:INN-hFSH-60
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Follicle Stimulating Hormone beta 2
Clonality: monoclonal
Clone: INN-hFSH-60
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human