Order Entry
Italy
ContactUsLinkComponent
Anti-FIBROBLAST GROWTH FACTOR-21 Rabbit polyclonal antibody
Anti-FIBROBLAST GROWTH FACTOR-21 Rabbit polyclonal antibody
Catalog # USBIF4212-93F
Supplier:  US Biological
Anti-FIBROBLAST GROWTH FACTOR-21 Rabbit polyclonal antibody
Catalog # USBIF4212-93F
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast Growth Factor-21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,2
  • Sequence:
    MRGSHHHHHHGMASHPIPDSSPLLQFGGQVRQRYLYTDDAQQTEAHLEIREDGTVGGAADQSPESLLQLKALKPGVIQILGVKTSRFLCQRPDGALYGSLHFDPEACSFRELLLEDGYNVYQSEAHGLPLHLPGNKSPHRDPAPRGPARFLPLPGLPPAPPEPPGILAPQPPDVGSSDPLSMVGPSQGRSPSYAS
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: FIBROBLAST GROWTH FACTOR-21
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human