Order Entry
Italy
ContactUsLinkComponent
Anti-Fibroblast Growth Factor 2, Basic, Neutralizing Mouse monoclonal antibody [clone: 0.T.50]
Anti-Fibroblast Growth Factor 2, Basic, Neutralizing Mouse monoclonal antibody [clone: 0.T.50]
Catalog # USBIF4210-22X
Supplier:  US Biological
Anti-Fibroblast Growth Factor 2, Basic, Neutralizing Mouse monoclonal antibody [clone: 0.T.50]
Catalog # USBIF4210-22X
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast Growth Factor 2, Basic, Neutralizing
  • Antigen symbol:
    FGF2
  • Clonality:
    Monoclonal
  • Clone:
    0.T.50
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Neutralisation:
    Yes
  • Form:
    liquid
  • Storage buffer:
    TBS
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation and DEAE-Sephacel chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Type: Primary
Antigen: Fibroblast Growth Factor 2, Basic, Neutralizing
Clonality: monoclonal
Clone: 0.T.50
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human, Mouse, Rat