Order Entry
Italy
ContactUsLinkComponent
 
Anti-Fibroblast Growth Factor 2, Basic Mouse monoclonal antibody [clone: 0.T.49]
  USBIF4210-21X
Anti-Fibroblast Growth Factor 2, Basic Mouse monoclonal antibody [clone: 0.T.49]
  USBIF4210-21X

 

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast Growth Factor 2, Basic
  • Antigen symbol:
    FGF2
  • Clonality:
    Monoclonal
  • Clone:
    0.T.49
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation and DEAE chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: Fibroblast Growth Factor 2, Basic
Clonality: monoclonal
Clone: 0.T.49
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human, Mouse, Rat