Order Entry
Italy
ContactUsLinkComponent
Anti-FK506 Binding Protien 51 Rabbit polyclonal antibody
Anti-FK506 Binding Protien 51 Rabbit polyclonal antibody
Catalog # USBIF4150-39
Supplier:  US Biological
Anti-FK506 Binding Protien 51 Rabbit polyclonal antibody
Catalog # USBIF4150-39
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FK506 binding protein 51
  • Antigen symbol:
    FKBP51
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Hamster,
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    394 to 407
  • Storage buffer:
    PBSwith 1mg/ml BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: FK506 Binding Protien 51
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Hamster, Human, Mouse, Rat