Order Entry
Italy
ContactUsLinkComponent
Anti-Estradiol, 17 beta Mouse Monoclonal Antibody [clone: 6F92]
Anti-Estradiol, 17 beta Mouse Monoclonal Antibody [clone: 6F92]
Catalog # USBIE3550-20
Supplier:  US Biological
Anti-Estradiol, 17 beta Mouse Monoclonal Antibody [clone: 6F92]
Catalog # USBIE3550-20
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Estradiol, 17 beta
  • Clonality:
    Monoclonal
  • Clone:
    6F92
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 15mM sodium azide, before the addition of glycerol to 40%.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Tested applications:
    Radio Immunoassay
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: Estradiol, 17 beta
Clonality: monoclonal
Clone: 6F92
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: