Order Entry
Italy
ContactUsLinkComponent
 
Anti-Endothelial Cell Antigen, Rat Mouse monoclonal antibody [clone: HIS-52 (RECA-1)]
Catalog # USBIE2291-90B
Supplier:  US Biological
Anti-Endothelial Cell Antigen, Rat Mouse monoclonal antibody [clone: HIS-52 (RECA-1)]
Catalog # USBIE2291-90B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endothelial Cell Antigen, Rat
  • Clonality:
    Monoclonal
  • Clone:
    HIS-52 (RECA-1)
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:40

Type: Primary
Antigen: Endothelial Cell Antigen, Rat
Clonality: monoclonal
Clone: HIS-52 (RECA-1)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat