Order Entry
Italy
ContactUsLinkComponent
Anti-DMP1 Goat Polyclonal Antibody
Anti-DMP1 Goat Polyclonal Antibody
  USBID8075-50B
Anti-DMP1 Goat Polyclonal Antibody
  USBID8075-50B

 

  • Antibody type:
    Primary
  • Antigen name:
    dentin matrix acidic phosphoprotein 1
  • Antigen symbol:
    DMP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
  • Sequence:
    C-ENSEQANTQDSG
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: ELISA: 1:4000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: DMP1 (dentin matrix acidic phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human