Order Entry
Italy
ContactUsLinkComponent
Anti-Cytochrome C Releasing Factor 21 Sheep Polyclonal Antibody
Anti-Cytochrome C Releasing Factor 21 Sheep Polyclonal Antibody
Catalog # USBIC9095-07A
Supplier:  US Biological
Anti-Cytochrome C Releasing Factor 21 Sheep Polyclonal Antibody
Catalog # USBIC9095-07A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome C Releasing Factor 21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Amino acid number:
    1 to 188
  • Storage buffer:
    0,2um filtered solution
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1.0 ?g/ml

Type: Primary
Antigen: Cytochrome C Releasing Factor 21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human