Order Entry
Italy
ContactUsLinkComponent
Anti-CBS Mouse Monoclonal Antibody [clone: 30]
Anti-CBS Mouse Monoclonal Antibody [clone: 30]
Catalog # USBIC8948-50D
Supplier:  US Biological
undefined
Anti-CBS Mouse Monoclonal Antibody [clone: 30]
Catalog # USBIC8948-50D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cystathionine-beta-synthase
  • Antigen symbol:
    CBS
  • Clonality:
    Monoclonal
  • Clone:
    30
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1 to 101
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Monoclonal
Clone: 30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human