Anti-CREB Rabbit polyclonal antibody
Catalog # USBIC7915-05D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CREB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Ser133
- Storage buffer:10mM HEPES, pH 7,5, 0,1% BSA, 0,09% BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot:1:1000
Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat