Order Entry
Italy
ContactUsLinkComponent
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 1.BB.98]
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 1.BB.98]
Catalog # USBIC7907-10-250
Supplier:  US Biological
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 1.BB.98]
Catalog # USBIC7907-10-250
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-Reactive Protein
  • Antigen symbol:
    CRP
  • Clonality:
    Monoclonal
  • Clone:
    1.BB.98
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Type: Primary
Antigen: C-Reactive Protein
Clonality: Monoclonal
Clone: 1.BB.98
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human