Order Entry
Italy
ContactUsLinkComponent
Anti-GJA1 Rabbit Polyclonal Antibody
Anti-GJA1 Rabbit Polyclonal Antibody
  USBIC7857-01H
Anti-GJA1 Rabbit Polyclonal Antibody
  USBIC7857-01H

 

  • Antibody type:
    Primary
  • Antigen name:
    gap junction Protein, alpha 1, 43kDa
  • Antigen symbol:
    GJA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 2?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: GJA1 (gap junction protein, alpha 1, 43kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat