Order Entry
Italy
ContactUsLinkComponent
Anti-Complement C9 Mouse Monoclonal Antibody [clone: 53]
Anti-Complement C9 Mouse Monoclonal Antibody [clone: 53]
Catalog # USBIC7850-53
Supplier:  US Biological
Anti-Complement C9 Mouse Monoclonal Antibody [clone: 53]
Catalog # USBIC7850-53
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complement C9
  • Antigen symbol:
    C9
  • Clonality:
    Monoclonal
  • Clone:
    53
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 1:20000

Type: Primary
Antigen: Complement C9
Clonality: Monoclonal
Clone: 53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human