Anti-Complement C3a, C3a des-Arg, Human Mouse Monoclonal Antibody [clone: 6F69]
Catalog # USBIC7850-13J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Complement C3a, C3a des-Arg, Human
- Clonality:Monoclonal
- Clone:6F69
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:0,01M PBS, pH 7,4, 15mM sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:32000
Type: Primary
Antigen: Complement C3a, C3a des-Arg, Human
Clonality: Monoclonal
Clone: 6F69
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human