Anti-CFL1 Rabbit monoclonal antibody [clone: 7H15]
Catalog # USBIC7506-20A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cofilin 1
- Antigen symbol:CFL1
- Clonality:Monoclonal
- Clone:7H15
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Modification:phospho-Ser3
- Storage buffer:10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, <0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:100
Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: Monoclonal
Clone: 7H15
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Monkey, Mouse, Rat