Anti-CFL1 Rabbit polyclonal antibody
USBIC7506-10A
- Antibody type:Primary
- Antigen name:Cofilin 1
- Antigen symbol:CFL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat