Order Entry
Italy
ContactUsLinkComponent
Anti-Clostridium sp. Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-Clostridium sp. Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIC5853-25A
Supplier:  US Biological
Anti-Clostridium sp. Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIC5853-25A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Clostridium sp.
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 10mg/ml BSA, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~4-5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: Clostridium sp.
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: