Order Entry
Italy
ContactUsLinkComponent
Anti-CEACAM 21 Rabbit polyclonal antibody
Anti-CEACAM 21 Rabbit polyclonal antibody
Catalog # USBIC2589-87P
Supplier:  US Biological
Anti-CEACAM 21 Rabbit polyclonal antibody
Catalog # USBIC2589-87P
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CEACAM 21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:500-1:1000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: CEACAM 21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human