Anti-IFNGR1 Mouse Monoclonal Antibody [clone: GIR-208]
USBIC2453-01J
- Antibody type:Primary
- Antigen name:Interferon gamma receptor 1
- Antigen symbol:IFNGR1
- Clonality:Monoclonal
- Clone:GIR-208
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: IFNGR1 (interferon gamma receptor 1)
Clonality: Monoclonal
Clone: GIR-208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human