Order Entry
Italy
ContactUsLinkComponent
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Catalog # USBIC2441-85H
Supplier:  US Biological
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Catalog # USBIC2441-85H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Thy1.2
  • Clonality:
    Monoclonal
  • Clone:
    30-H12
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS,sodium azide, stabilizing agent.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: Thy1.2 (Thy1.2)
Clonality: Monoclonal
Clone: 30-H12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse