Order Entry
Italy
ContactUsLinkComponent
Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: F15-42-1]
Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: F15-42-1]
Catalog # USBIC2441-02F3
Supplier:  US Biological
Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: F15-42-1]
Catalog # USBIC2441-02F3
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thy1 Cell Surface Antigen
  • Antigen symbol:
    THY1
  • Clonality:
    Monoclonal
  • Clone:
    F15-42-1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:10-1:15
Do not freeze!

Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: