Order Entry
Italy
ContactUsLinkComponent
 
Anti-CD59 Glycoprotein Mouse Monoclonal Antibody [clone: MEM-43/5]
  USBIC2413-22C
Anti-CD59 Glycoprotein Mouse Monoclonal Antibody [clone: MEM-43/5]
  USBIC2413-22C

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD59 Glycoprotein
  • Clonality:
    Monoclonal
  • Clone:
    MEM-43/5
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Flow Cytometry: 1 ?g/ml; Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CD59 Glycoprotein
Clonality: Monoclonal
Clone: MEM-43/5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human