Order Entry
Italy
ContactUsLinkComponent
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 6-13-19-1]
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 6-13-19-1]
Catalog # USBIC2411-22H
Supplier:  US Biological
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 6-13-19-1]
Catalog # USBIC2411-22H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P)
  • Antigen symbol:
    B3GAT1
  • Clonality:
    Monoclonal
  • Clone:
    6-13-19-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Format:
    ascites
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:5

Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: Monoclonal
Clone: 6-13-19-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human