Anti-NCAM1 Mouse Monoclonal Antibody [clone: 12C3]
Catalog # USBIC2410-60J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Neural Cell Adhesion Molecule 1
- Antigen symbol:NCAM1
- Clonality:Monoclonal
- Clone:12C3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 1:10
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 12C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: