Anti-CD53 Mouse Monoclonal Antibody [clone: MEM-53]
Catalog # USBIC2408-12D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD53 molecule
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:MEM-53
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10-1:25 Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CD53 (CD53 molecule)
Clonality: Monoclonal
Clone: MEM-53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human