Order Entry
Italy
ContactUsLinkComponent
Anti-ITGA5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5H10-27 (MFR5)]
Anti-ITGA5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5H10-27 (MFR5)]
Catalog # USBIC2405-02E
Supplier:  US Biological
Anti-ITGA5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5H10-27 (MFR5)]
Catalog # USBIC2405-02E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin Alpha 5
  • Antigen symbol:
    ITGA5
  • Clonality:
    Monoclonal
  • Clone:
    5H10-27 (MFR5)
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide and a stabilizing agent.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: Monoclonal
Clone: 5H10-27 (MFR5)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse