Anti-CD45.2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 104-2]
USBIC2400-91N
- Antibody type:Primary
- Antigen symbol:CD45.2
- Clonality:Monoclonal
- Clone:104-2
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Do not freeze!
Type: Primary
Antigen: CD45.2 (CD45.2)
Clonality: Monoclonal
Clone: 104-2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse