Order Entry
Italy
ContactUsLinkComponent
 
Anti-CD45, Mixed Clones Mouse Monoclonal Antibody [clone: PD7/26/16+2B11]
Catalog # USBIC2399-09A
Supplier:  US Biological
Anti-CD45, Mixed Clones Mouse Monoclonal Antibody [clone: PD7/26/16+2B11]
Catalog # USBIC2399-09A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    PD7/26/16+2B11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    10mM PBS, pH 7,4, 0,2% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CD45, Mixed Clones
Clonality: Monoclonal
Clone: PD7/26/16+2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human