Order Entry
Italy
ContactUsLinkComponent
Anti-SPN Mouse Monoclonal Antibody [clone: MEM-59]
Anti-SPN Mouse Monoclonal Antibody [clone: MEM-59]
Catalog # USBIC2397-76S
Supplier:  US Biological
Anti-SPN Mouse Monoclonal Antibody [clone: MEM-59]
Catalog # USBIC2397-76S
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sialophorin
  • Antigen symbol:
    SPN
  • Clonality:
    Monoclonal
  • Clone:
    MEM-59
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Donkey,
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: SPN (sialophorin)
Clonality: Monoclonal
Clone: MEM-59
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Burro, Human