Anti-FUT4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K229]
Catalog # USBIC2266-01F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fucosyltransferase 4
- Antigen symbol:FUT4
- Clonality:Monoclonal
- Clone:5K229
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~100tests/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: Monoclonal
Clone: 5K229
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human