Order Entry
Italy
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-18]
Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-18]
Catalog # USBIC2265-49C
Supplier:  US Biological
Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-18]
Catalog # USBIC2265-49C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD14 molecule
  • Antigen symbol:
    CD14
  • Clonality:
    Monoclonal
  • Clone:
    MEM-18
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by precipitation method
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: MEM-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human