Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 53-6.7]
USBIC2259-97A3
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:PE (Phycoerythrin)/Cy5®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Recommended dilution: Flow Cytometry:1:5;
Do not freeze!
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse