Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-6.7]
Catalog # USBIC2259-28K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse