Anti-CD5 Rat Monoclonal Antibody (Biotin) [clone: 5K5]
USBIC2256-23B
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:5K5
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: Monoclonal
Clone: 5K5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse