Anti-CD5 Mouse Monoclonal Antibody (Biotin) [clone: 5K207]
Catalog # USBIC2255-85K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:5K207
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: Monoclonal
Clone: 5K207
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human