Order Entry
Italy
ContactUsLinkComponent
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-57]
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-57]
Catalog # USBIC2254-56D2
Supplier:  US Biological
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-57]
Catalog # USBIC2254-56D2
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD3
  • Clonality:
    Monoclonal
  • Clone:
    MEM-57
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS,BSA, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Recommended dilution: Flow Cytometry: 20?l reagent/100?l whole blood
Do not freeze!

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: MEM-57
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human